missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals Product Code.: 15475416

1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals

Product Code. 15475416
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15475416

Brand: Thermo Scientific Alfa Aesar H60017.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528854-34-4
Molecular Formula C22H44P2
Molecular Weight (g/mol) 370.542
MDL Number MFCD08705244
InChI Key IRVIFEWWPYKALC-GXRSIYKFSA-N
Synonym 1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan
PubChem CID 11707202
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane
SMILES CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C

Specifications

Melting Point 67°C to 72°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 370.53
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC26Z036 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60017.03,H60017.MD
Lot NumberY12F022 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60017.03,H60017.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.