missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylphosphino)ethane, 97%, Thermo Scientific Chemicals

50.80 € - 151.00 €

Chemical Identifiers

CAS 23936-60-9
Molecular Formula C6H16P2
Molecular Weight (g/mol) 150.14
MDL Number MFCD00008511
InChI Key ZKWQSBFSGZJNFP-UHFFFAOYSA-N
Synonym 1,2-bis dimethylphosphino ethane, ethylenebis dimethylphosphine, phosphine, 1,2-ethanediylbis dimethyl, 2-dimethylphosphanylethyl dimethyl phosphane, dimethyl-2-dimethylphosphino ethylphosphine, 2-dimethylphosphanyl ethyl dimethylphosphane, pubchem6482, acmc-1cjp0, ghl.pd_mitscher_leg0.173, zkwqsbfsgzjnfp-uhfffaoysa
PubChem CID 141059
IUPAC Name 2-dimethylphosphanylethyl(dimethyl)phosphane
SMILES CP(C)CCP(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21051.01
0.25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11440147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21051.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23936-60-9
150.14
ZKWQSBFSGZJNFP-UHFFFAOYSA-N
141059
CP(C)CCP(C)C
C6H16P2
MFCD00008511
1,2-bis dimethylphosphino ethane, ethylenebis dimethylphosphine, phosphine, 1,2-ethanediylbis dimethyl, 2-dimethylphosphanylethyl dimethyl phosphane, dimethyl-2-dimethylphosphino ethylphosphine, 2-dimethylphosphanyl ethyl dimethylphosphane, pubchem6482, acmc-1cjp0, ghl.pd_mitscher_leg0.173, zkwqsbfsgzjnfp-uhfffaoysa
2-dimethylphosphanylethyl(dimethyl)phosphane

Specifications

23936-60-9
0.9
−17°C (1.4°F)
C6H16P2
1.507
UN1993
1,2-bis dimethylphosphino ethane, ethylenebis dimethylphosphine, phosphine, 1,2-ethanediylbis dimethyl, 2-dimethylphosphanylethyl dimethyl phosphane, dimethyl-2-dimethylphosphino ethylphosphine, 2-dimethylphosphanyl ethyl dimethylphosphane, pubchem6482, acmc-1cjp0, ghl.pd_mitscher_leg0.173, zkwqsbfsgzjnfp-uhfffaoysa
CP(C)CCP(C)C
150.14
150.14
1,2-Bis(dimethylphosphino)ethane
179°C to 180°C
180°C
Recognizable
(CH3)2PCH2CH2P(CH3)2
MFCD00008511
Air Sensitive
ZKWQSBFSGZJNFP-UHFFFAOYSA-N
2-dimethylphosphanylethyl(dimethyl)phosphane
141059
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P222-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H225-H250-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: I

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only