missing translation for 'onlineSavingsMsg'
Learn More
1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals Product Code.: 13498588

1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals

Product Code. 13498588
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13498588

Brand: Thermo Scientific Acros 445641000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, clorobenShow MoreShow Less
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point -15.0°C
Density 1.3000g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 67°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Cl2
Refractive Index 1.5500 to 1.5520
Quantity 100 mL
Merck Index 15,3065
Specific Gravity 1.3
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Formula Weight 147
Percent Purity 99+%
Grade HPLC
Chemical Name or Material 1, 2-Dichlorobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464928 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number445640010,445641000
Lot NumberA0462637 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number445640010,445641000
Lot NumberA0457191 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number445640010,445641000
Lot NumberA0460682 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number445640010,445641000
Lot NumberA0457969 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number445640010,445641000
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : o-Dichlorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.