Learn More
1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals
19.82 € - 542.70 €
Chemical Identifiers
CAS | 95-50-1 |
---|---|
MDL Number | MFCD00000535 |
InChI Key | RFFLAFLAYFXFSW-UHFFFAOYSA-N |
Synonym | o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben |
PubChem CID | 7239 |
ChEBI | CHEBI:35290 |
IUPAC Name | 1,2-dichlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10396170
|
Thermo Scientific Acros
113180050 |
5 mL | Glass Bottle | N/A | |||||
10530371
|
Thermo Scientific Acros
113180010 |
1 L | Glass Bottle |
48.54 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10060900
|
Thermo Scientific Acros
113180025 |
2.5 L | Glass Bottle |
93.26 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10751734
|
Thermo Scientific Acros
113180250 |
25 L | Metal drum |
542.70 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-50-1 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
Specifications
95-50-1 | |
100.0 | |
1.3000g/mL | |
99% | |
C6H4Cl2 | |
MFCD00000535 | |
15, 3065 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
≥98.5% | |
Liquid |
98.5 | |
Colorless to Yellow | |
Authentic | |
C6H4Cl2 | |
1.5500 to 1.5520 (20°C, 589nm) | |
05, 201 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
C1=CC=C(C(=C1)Cl)Cl | |
146.998 | |
CHEBI:35290 | |
1.3mbar at 20°C | |
Pure | |
1, 2-Dichlorobenzene |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only