missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[2-Methoxy-5-(trifluoromethoxy)phenyl]ethanol, 95%, Thermo Scientific™
87.74 € - 263.00 €
Chemical Identifiers
CAS | 468074-91-1 |
---|---|
Molecular Formula | C10H11F3O3 |
Molecular Weight (g/mol) | 236.19 |
MDL Number | MFCD08062374 |
InChI Key | DKUPMKWSJITPFC-UHFFFAOYNA-N |
Synonym | 1-2-methoxy-5-trifluoromethoxy phenyl ethan-1-ol, 1-2-methoxy-5-trifluoromethoxy phenyl ethanol, 1-hydroxy-1-2-methoxy-5-triflouromethoxy phenyl ethane |
PubChem CID | 18542515 |
IUPAC Name | 1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethanol |
SMILES | COC1=C(C=C(OC(F)(F)F)C=C1)C(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497885
|
Thermo Scientific Alfa Aesar
H31765.03 |
1 g |
87.74 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407895
|
Thermo Scientific Alfa Aesar
H31765.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
468074-91-1 | |
236.19 | |
DKUPMKWSJITPFC-UHFFFAOYNA-N | |
18542515 | |
COC1=C(C=C(OC(F)(F)F)C=C1)C(C)O |
C10H11F3O3 | |
MFCD08062374 | |
1-2-methoxy-5-trifluoromethoxy phenyl ethan-1-ol, 1-2-methoxy-5-trifluoromethoxy phenyl ethanol, 1-hydroxy-1-2-methoxy-5-triflouromethoxy phenyl ethane | |
1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethanol |
Specifications
468074-91-1 | |
MFCD08062374 | |
DKUPMKWSJITPFC-UHFFFAOYNA-N | |
1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethanol | |
18542515 | |
95% |
C10H11F3O3 | |
1-2-methoxy-5-trifluoromethoxy phenyl ethan-1-ol, 1-2-methoxy-5-trifluoromethoxy phenyl ethanol, 1-hydroxy-1-2-methoxy-5-triflouromethoxy phenyl ethane | |
COC1=C(C=C(OC(F)(F)F)C=C1)C(C)O | |
236.19 | |
236.19 | |
1-[2-Methoxy-5-(trifluoromethoxy)phenyl]ethanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures