missing translation for 'onlineSavingsMsg'
Learn More
1,2-Methylenedioxy-4-nitrobenzene, 98+%, Thermo Scientific Chemicals Product Code.: 11476616

1,2-Methylenedioxy-4-nitrobenzene, 98+%, Thermo Scientific Chemicals

Product Code. 11476616
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11476616

Brand: Thermo Scientific Alfa Aesar B24838.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-44-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00005824
InChI Key SNWQAKNKGGOVMO-UHFFFAOYSA-N
Synonym 5-nitrobenzo d 1,3 dioxole, 1,2-methylenedioxy-4-nitrobenzene, 3,4-methylenedioxynitrobenzene, 1,3-benzodioxole, 5-nitro, methylenedioxynitrobenzene, 5-nitrobenzodioxole, 3,4-methylenedioxy-1-nitrobenzene, 5-nitro-2h-1,3-benzodioxole, benzene, 1,2-methylenedioxy-4-nitroShow MoreShow Less
PubChem CID 75798
IUPAC Name 5-nitro-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 142°C to 149°C
Flash Point 142°C (287°F)
Quantity 5 g
Beilstein 169063
Formula Weight 167.12
Percent Purity ≥98%
Chemical Name or Material 1,2-Methylenedioxy-4-nitrobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ8526A Certificate TypeCertificate of Analysis Date12/03/2024 Catalog NumberB24838.06
Lot NumberH8315A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24838.06,B24838.14,B24838.22
Lot NumberE3911A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24838.06,B24838.14,B24838.22
Lot NumberP03809 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24838.06,B24838.14,B24838.22
Lot NumberH7112A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24838.06,B24838.14,B24838.22
compliance-icons
Product Identifier
  • 1,2-Methylenedioxy-4-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Methylenedioxy-4-nitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Methylenedioxy-4-nitrobenzene, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.