missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in dimethyl sulfoxide (74 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), water (<1mg/mL at 25°C), and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Soluble in dimethyl sulfoxide (74 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), water (<1mg/mL at 25°C), and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.

Specifications
Specifications
CAS | 633-65-8 |
Molecular Formula | C20H18ClNO4 |
MDL Number | MFCD00011939 |
Quantity | 5g |
Beilstein | 3836585 |
Synonym | berberine chloride, berberine hydrochloride, natural yellow 18, benzodioxide, berberinium chloride, berberine hcl, unii-uot4o1byv8, berberine chloride jan, uot4o1byv8 |
Solubility Information | Soluble in dimethyl sulfoxide (74mg/ml at 25°C),ethanol (<1mg/ml at 25°C),water (<1mg/ml at 25°C),and methanol. |
InChI Key | VKJGBAJNNALVAV-UHFFFAOYSA-M |
SMILES | COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.[Cl-] |
Molecular Weight (g/mol) | 371.817 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T01K031 | Certificate of Analysis | 25/07/2024 | J62311.06 |
Q29K532 | Certificate of Analysis | 25/07/2024 | J62311.06 |
X28J525 | Certificate of Analysis | 14/05/2024 | J62311.06 |
R19J559 | Certificate of Analysis | 09/08/2023 | J62311.06 |
Q27I064 | Certificate of Analysis | 17/11/2022 | J62311.06 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Berberine chloride, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction