Learn More
1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals
46.80 € - 163.00 €
Chemical Identifiers
CAS | 102-04-5 |
---|---|
Molecular Formula | C15H14O |
Molecular Weight (g/mol) | 210.28 |
MDL Number | MFCD00004795 |
InChI Key | YFKBXYGUSOXJGS-UHFFFAOYSA-N |
Synonym | 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 |
PubChem CID | 7593 |
IUPAC Name | 1,3-diphenylpropan-2-one |
SMILES | O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387855
|
Thermo Scientific Alfa Aesar
L02830.14 |
25 g |
46.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397855
|
Thermo Scientific Alfa Aesar
L02830.22 |
100 g |
163.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
102-04-5 | |
210.28 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
7593 | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
C15H14O | |
MFCD00004795 | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
1,3-diphenylpropan-2-one |
Specifications
102-04-5 | |
1.04 | |
>110°C (230°F) | |
MFCD00004795 | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
1,3-diphenylpropan-2-one | |
7593 | |
≥98% |
32°C to 36°C | |
330°C | |
C15H14O | |
974767 | |
Slightly soluble in water. | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
210.28 | |
210.28 | |
1,3-Diphenylacetone |
Safety and Handling
missing translation for 'einecsNumber' : 203-000-0
missing translation for 'rtecsNumber' : UC2010030
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only