missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(Trifluoromethoxy)phenyl]ethanol, 97%, Thermo Scientific™
49.40 € - 154.00 €
Chemical Identifiers
CAS | 347194-02-9 |
---|---|
Molecular Formula | C9H9F3O2 |
Molecular Weight (g/mol) | 206.164 |
MDL Number | MFCD06797343 |
InChI Key | BWANKNAGTRWECQ-UHFFFAOYSA-N |
Synonym | 1-3-trifluoromethoxy phenyl ethanol, 1-3-trifluoromethoxy phenyl ethan-1-ol, 1-3-trifluoromethoxyphenyl ethanol |
PubChem CID | 44891072 |
IUPAC Name | 1-[3-(trifluoromethoxy)phenyl]ethanol |
SMILES | CC(C1=CC(=CC=C1)OC(F)(F)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495107
|
Thermo Scientific Alfa Aesar
H32390.06 |
5 g |
49.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405117
|
Thermo Scientific Alfa Aesar
H32390.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
347194-02-9 | |
206.164 | |
BWANKNAGTRWECQ-UHFFFAOYSA-N | |
44891072 | |
CC(C1=CC(=CC=C1)OC(F)(F)F)O |
C9H9F3O2 | |
MFCD06797343 | |
1-3-trifluoromethoxy phenyl ethanol, 1-3-trifluoromethoxy phenyl ethan-1-ol, 1-3-trifluoromethoxyphenyl ethanol | |
1-[3-(trifluoromethoxy)phenyl]ethanol |
Specifications
347194-02-9 | |
MFCD06797343 | |
BWANKNAGTRWECQ-UHFFFAOYSA-N | |
1-[3-(trifluoromethoxy)phenyl]ethanol | |
44891072 | |
97% |
C9H9F3O2 | |
1-3-trifluoromethoxy phenyl ethanol, 1-3-trifluoromethoxy phenyl ethan-1-ol, 1-3-trifluoromethoxyphenyl ethanol | |
CC(C1=CC(=CC=C1)OC(F)(F)F)O | |
206.164 | |
206.16 | |
1-[3-(Trifluoromethoxy)phenyl]ethanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures