Learn More
1-(4-Biphenylyl)ethanol, 98%, Thermo Scientific Chemicals
44.90 € - 155.00 €
Chemical Identifiers
CAS | 3562-73-0 |
---|---|
Molecular Formula | C14H14O |
Molecular Weight (g/mol) | 198.265 |
MDL Number | MFCD00016859 |
InChI Key | GOISDOCZKZYADO-UHFFFAOYSA-N |
Synonym | 1-4-biphenylyl ethanol, 4-1-hydroxyethyl biphenyl, 1-biphenyl-4-yl ethanol, diascleril, difencol, drucoles, 1-4-phenylphenyl ethanol, 1-4-biphenyl ethanol, 4-biphenylmethanol, .alpha.-methyl, 1-1,1'-biphenyl-4-yl ethanol |
PubChem CID | 96176 |
IUPAC Name | 1-(4-phenylphenyl)ethanol |
SMILES | CC(C1=CC=C(C=C1)C2=CC=CC=C2)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3562-73-0 | |
198.265 | |
GOISDOCZKZYADO-UHFFFAOYSA-N | |
96176 | |
CC(C1=CC=C(C=C1)C2=CC=CC=C2)O |
C14H14O | |
MFCD00016859 | |
1-4-biphenylyl ethanol, 4-1-hydroxyethyl biphenyl, 1-biphenyl-4-yl ethanol, diascleril, difencol, drucoles, 1-4-phenylphenyl ethanol, 1-4-biphenyl ethanol, 4-biphenylmethanol, .alpha.-methyl, 1-1,1'-biphenyl-4-yl ethanol | |
1-(4-phenylphenyl)ethanol |
Specifications
3562-73-0 | |
C14H14O | |
2044710 | |
GOISDOCZKZYADO-UHFFFAOYSA-N | |
1-(4-phenylphenyl)ethanol | |
96176 | |
98% |
94°C to 99°C | |
MFCD00016859 | |
1-4-biphenylyl ethanol, 4-1-hydroxyethyl biphenyl, 1-biphenyl-4-yl ethanol, diascleril, difencol, drucoles, 1-4-phenylphenyl ethanol, 1-4-biphenyl ethanol, 4-biphenylmethanol, .alpha.-methyl, 1-1,1'-biphenyl-4-yl ethanol | |
CC(C1=CC=C(C=C1)C2=CC=CC=C2)O | |
198.265 | |
198.27 | |
1-(4-Biphenylyl)ethanol |
Safety and Handling
missing translation for 'einecsNumber' : 222-629-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.