Learn More
1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals
45.12 € - 154.77 €
Chemical Identifiers
CAS | 280-57-9 |
---|---|
Molecular Formula | C6H12N2 |
Molecular Weight (g/mol) | 112.176 |
MDL Number | MFCD00006689 |
InChI Key | IMNIMPAHZVJRPE-UHFFFAOYSA-N |
Synonym | 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25Show More |
PubChem CID | 9237 |
IUPAC Name | 1,4-diazabicyclo[2.2.2]octane |
SMILES | C1CN2CCN1CC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497856
|
Thermo Scientific Alfa Aesar
A14003.22 |
100 g |
45.12 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407866
|
Thermo Scientific Alfa Aesar
A14003.36 |
500 g |
154.77 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
280-57-9 | |
112.176 | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
9237 | |
C1CN2CCN1CC2 |
C6H12N2 | |
MFCD00006689 | |
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25Show More | |
1,4-diazabicyclo[2.2.2]octane |
Specifications
280-57-9 | |
1.14 | |
62°C (143°F) | |
C6H12N2 | |
UN1325 | |
Hygroscopic | |
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
1,4-diazabicyclo[2.2.2]octane | |
9237 | |
98% |
155°C to 160°C | |
174°C to 176°C | |
Amine-like | |
MFCD00006689 | |
103618 | |
14,9669 | |
Soluble in water,acetone,benzene,ethanol and methyl ethyl ketone. | |
C1CN2CCN1CC2 | |
112.176 | |
112.17 | |
1,4-Diazabicyclo[2.2.2]octane |
Safety and Handling
GHS H Statement
H228-H318-H302-H335-H315
Flammable solid.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H318-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 205-999-9
missing translation for 'rtecsNumber' : HM0354200
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1,4-Diazabicyclo¢2.2.2!octane
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.