Learn More
1,5-Diaminopentane, 98%, Thermo Scientific Chemicals
97.69 € - 412.05 €
Chemical Identifiers
CAS | 462-94-2 |
---|---|
Molecular Formula | C5H14N2 |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00008239 |
InChI Key | VHRGRCVQAFMJIZ-UHFFFAOYSA-N |
Synonym | 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p |
PubChem CID | 273 |
ChEBI | CHEBI:18127 |
IUPAC Name | pentane-1,5-diamine |
SMILES | NCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456226
|
Thermo Scientific Alfa Aesar
B23039.06 |
5 g |
97.69 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466226
|
Thermo Scientific Alfa Aesar
B23039.14 |
25 g |
412.05 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,5-Diaminopentane is mainly used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Diaminopentane is mainly used in organic synthesis.
Solubility
Soluble in water.
Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
Chemical Identifiers
462-94-2 | |
102.18 | |
VHRGRCVQAFMJIZ-UHFFFAOYSA-N | |
273 | |
pentane-1,5-diamine |
C5H14N2 | |
MFCD00008239 | |
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p | |
CHEBI:18127 | |
NCCCCCN |
Specifications
462-94-2 | |
0.873 | |
0.98 | |
H2N(CH2)5NH2 | |
MFCD00008239 | |
Air sensitive; Hygroscopic | |
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p | |
VHRGRCVQAFMJIZ-UHFFFAOYSA-N | |
pentane-1,5-diamine | |
273 | |
102.18 | |
Liquid |
9°C | |
178°C to 180°C | |
C5H14N2 | |
1.4585 | |
UN2735 | |
14,1609 | |
Soluble in water. | |
NCCCCCN | |
102.18 | |
CHEBI:18127 | |
98% | |
1, 5-Diaminopentane |
Safety and Handling
GHS H Statement
H314-H318-H227
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 207-329-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 1,5-Diaminopentane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Pentamethylenediamine
RUO – Research Use Only