missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2-methoxyphenyl)ethanol, 95%, Thermo Scientific™

54.20 € - 494.00 €

Chemical Identifiers

CAS 878572-08-8
Molecular Formula C9H11FO2
Molecular Weight (g/mol) 170.18
MDL Number MFCD08062369
InChI Key VLBOHPCLXJIASU-UHFFFAOYNA-N
Synonym 1-5-fluoro-2-methoxyphenyl ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol, benzenemethanol,5-fluoro-2-methoxy-a-methyl, 1-5-fluoro-2-methoxy-phenyl-ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol_x000d_
PubChem CID 44891074
IUPAC Name 1-(5-fluoro-2-methoxyphenyl)ethanol
SMILES COC1=C(C=C(F)C=C1)C(C)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424697
View Documents
Thermo Scientific Alfa Aesar
H32022.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15404697
View Documents
Thermo Scientific Alfa Aesar
H32022.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15414697
View Documents
Thermo Scientific Alfa Aesar
H32022.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

878572-08-8
170.18
VLBOHPCLXJIASU-UHFFFAOYNA-N
44891074
COC1=C(C=C(F)C=C1)C(C)O
C9H11FO2
MFCD08062369
1-5-fluoro-2-methoxyphenyl ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol, benzenemethanol,5-fluoro-2-methoxy-a-methyl, 1-5-fluoro-2-methoxy-phenyl-ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol_x000d_
1-(5-fluoro-2-methoxyphenyl)ethanol

Specifications

878572-08-8
MFCD08062369
VLBOHPCLXJIASU-UHFFFAOYNA-N
1-(5-fluoro-2-methoxyphenyl)ethanol
44891074
95%
C9H11FO2
1-5-fluoro-2-methoxyphenyl ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol, benzenemethanol,5-fluoro-2-methoxy-a-methyl, 1-5-fluoro-2-methoxy-phenyl-ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol_x000d_
COC1=C(C=C(F)C=C1)C(C)O
170.18
170.18
1-(5-Fluoro-2-methoxyphenyl)ethanol
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.