Learn More
1-(6-chloro-1,3-benzothiazol-2-yl)hydrazine, 97%, Thermo Scientific™
69.20 € - 435.00 €
Chemical Identifiers
CAS | 51011-54-2 |
---|---|
Molecular Formula | C7H6ClN3S |
Molecular Weight (g/mol) | 199.656 |
MDL Number | MFCD00832729 |
InChI Key | JBAIRAQEQLLZMN-UHFFFAOYSA-N |
Synonym | 6-chloro-2-hydrazinylbenzo d thiazole, 1-6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-benzothiazol-2-yl-hydrazine, 6-chloro-2-hydrazinyl-1,3-benzothiazole, 1-6-chlorobenzo d thiazol-2-yl hydrazine, 6-chloro-2-hydrazino-1,3-benzothiazole, 6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-2-hydrazinobenzothiazole, 2-hydrazino-6-chloro-benzothiazole, 6-chlorobenzothiazole-2-ylhydrazine |
PubChem CID | 737197 |
IUPAC Name | (6-chloro-1,3-benzothiazol-2-yl)hydrazine |
SMILES | C1=CC2=C(C=C1Cl)SC(=N2)NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10703073
|
Thermo Scientific Maybridge
SEW01279CB |
250 mg |
69.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10465902
|
Thermo Scientific Maybridge
SEW01279DA |
1 g |
119.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10526785
|
Thermo Scientific Maybridge
SEW01279DE |
5 g |
435.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
51011-54-2 | |
199.656 | |
JBAIRAQEQLLZMN-UHFFFAOYSA-N | |
737197 | |
C1=CC2=C(C=C1Cl)SC(=N2)NN |
C7H6ClN3S | |
MFCD00832729 | |
6-chloro-2-hydrazinylbenzo d thiazole, 1-6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-benzothiazol-2-yl-hydrazine, 6-chloro-2-hydrazinyl-1,3-benzothiazole, 1-6-chlorobenzo d thiazol-2-yl hydrazine, 6-chloro-2-hydrazino-1,3-benzothiazole, 6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-2-hydrazinobenzothiazole, 2-hydrazino-6-chloro-benzothiazole, 6-chlorobenzothiazole-2-ylhydrazine | |
(6-chloro-1,3-benzothiazol-2-yl)hydrazine |
Specifications
51011-54-2 | |
97% | |
C7H6ClN3S | |
6-chloro-2-hydrazinylbenzo d thiazole, 1-6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-benzothiazol-2-yl-hydrazine, 6-chloro-2-hydrazinyl-1,3-benzothiazole, 1-6-chlorobenzo d thiazol-2-yl hydrazine, 6-chloro-2-hydrazino-1,3-benzothiazole, 6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-2-hydrazinobenzothiazole, 2-hydrazino-6-chloro-benzothiazole, 6-chlorobenzothiazole-2-ylhydrazine | |
C1=CC2=C(C=C1Cl)SC(=N2)NN | |
199.656 | |
199.66 | |
1-(6-Chloro-1, 3-benzothiazol-2-yl)hydrazine |
97.0 | |
Amber glass bottle | |
MFCD00832729 | |
JBAIRAQEQLLZMN-UHFFFAOYSA-N | |
(6-chloro-1,3-benzothiazol-2-yl)hydrazine | |
737197 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning