missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiyne, 98%, Thermo Scientific Chemicals

85.22 € - 291.45 €

Chemical Identifiers

CAS 871-84-1
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008580
InChI Key DSOJWVLXZNRKCS-UHFFFAOYSA-N
Synonym 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 70099
IUPAC Name octa-1,7-diyne
SMILES C#CCCCCC#C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10772211
View Documents Promotion Details
Thermo Scientific Acros
171340100
10 g Glass bottle
85.22 €
10g
Please to purchase this item. Need a web account? Register with us today!
10002100
View Documents Promotion Details
Thermo Scientific Acros
171340500
50 g Glass bottle
291.45 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

871-84-1
106.17
DSOJWVLXZNRKCS-UHFFFAOYSA-N
70099
C#CCCCCC#C
C8H10
MFCD00008580
1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci
octa-1,7-diyne

Specifications

871-84-1
100.0
0.8000g/mL
23°C
97.5% min. (GC)
HC≡C(CH2)4C≡CH
MFCD00008580
0.8
DSOJWVLXZNRKCS-UHFFFAOYSA-N
octa-1,7-diyne
70099
98%
1, 7-Octadiyne, 98%
98.5
Colorless to Yellow
135.0°C to 136.0°C
Authentic
C8H10
1.4454 to 1.4474
01, II, 248
1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci
C#CCCCCC#C
106.17
106.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 212-815-0

missing translation for 'tsca' : TSCA

Product Identifier
  • 1,7-Octadiyne

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only