missing translation for 'onlineSavingsMsg'
Learn More
1,7-Octadiyne, 98%, Thermo Scientific Chemicals Product Code.: 10772211

1,7-Octadiyne, 98%, Thermo Scientific Chemicals

Product Code. 10772211
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10772211

Brand: Thermo Scientific Acros 171340100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871-84-1
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008580
InChI Key DSOJWVLXZNRKCS-UHFFFAOYSA-N
Synonym 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ciShow MoreShow Less
PubChem CID 70099
IUPAC Name octa-1,7-diyne
SMILES C#CCCCCC#C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 135.0°C to 136.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡C(CH2)4C≡CH
Refractive Index 1.4454 to 1.4474
Quantity 10 g
Beilstein 01, II, 248
Specific Gravity 0.8
Formula Weight 106.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 7-Octadiyne, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452688 Certificate TypeCertificate of Analysis Date28/04/2023 Catalog Number171340500,171340100
Lot NumberA0401799 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number171340500,171340100
Lot NumberA0394523 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number171340500,171340100
Lot NumberA0444329 Certificate TypeCertificate of Analysis Date01/08/2022 Catalog Number171340500,171340100
Lot NumberA0432581 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number171340500,171340100
compliance-icons
Product Identifier
  • 1,7-Octadiyne
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,7-Octadiyne, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.