Learn More
1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals
24.92 € - 342.71 €
Chemical Identifiers
CAS | 1143-38-0 |
---|---|
Molecular Formula | C14H10O3 |
Molecular Weight (g/mol) | 226.23 |
MDL Number | MFCD00053409 |
InChI Key | NUZWLKWWNNJHPT-UHFFFAOYSA-N |
Synonym | anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy |
PubChem CID | 2202 |
ChEBI | CHEBI:37510 |
IUPAC Name | 1,8-dihydroxy-10H-anthracen-9-one |
SMILES | OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15505276
|
Thermo Scientific Acros
457770010 |
1 g |
24.92 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15560311
|
Thermo Scientific Acros
457770050 |
5 g |
89.14 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15515276
|
Thermo Scientific Acros
457770250 |
25 g |
342.71 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1143-38-0 | |
226.23 | |
NUZWLKWWNNJHPT-UHFFFAOYSA-N | |
2202 | |
1,8-dihydroxy-10H-anthracen-9-one |
C14H10O3 | |
MFCD00053409 | |
anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy | |
CHEBI:37510 | |
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 |
Specifications
1143-38-0 | |
Authentic | |
Glass Bottle | |
MFCD00053409 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform, acetone, benzene, slightly soluble in alcohol, ether, glacial acetic | |
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 | |
226.23 | |
CHEBI:37510 | |
97% |
178.0°C to 181.0°C | |
96% min. (HPLC) | |
C14H10O3 | |
anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy | |
NUZWLKWWNNJHPT-UHFFFAOYSA-N | |
1,8-dihydroxy-10H-anthracen-9-one | |
2202 | |
226.23 | |
1,8,9-Trihydroxyanthracene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-538-
- 1,8,9-Trihydroxyanthracene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1,8-Dihydroxyanthracen-9(10H)-one
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.