missing translation for 'onlineSavingsMsg'
Learn More
1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals Product Code.: 15505276

1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals

Product Code. 15505276
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15505276

Brand: Thermo Scientific Acros 457770010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
Synonym anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxyShow MoreShow Less
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-10H-anthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

Melting Point 178.0°C to 181.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, acetone, benzene, slightly soluble in alcohol, ether, glacial acetic
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 1,8,9-Trihydroxyanthracene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464110 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number457770250,457770050,457770010
Lot NumberA0452848 Certificate TypeCertificate of Analysis Date08/05/2023 Catalog Number457770250,457770050,457770010
Lot NumberA0409823 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number457770250,457770050,457770010
Lot NumberA0416126 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number457770250,457770050,457770010
compliance-icons
Product Identifier
  • 1,8,9-Trihydroxyanthracene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,8-Dihydroxyanthracen-9(10H)-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.