missing translation for 'onlineSavingsMsg'
Learn More
1,8-Cineole, 99%, Thermo Scientific Chemicals Product Code.: 10695522

1,8-Cineole, 99%, Thermo Scientific Chemicals

Product Code. 10695522
5 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

Product Code. 10695522

Brand: Thermo Scientific Acros 110340050

Please to purchase this item. Need a web account? Register with us today!

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 1.5°C
Color Colorless to Yellow
Density 0.9225g/mL
Boiling Point 176°C to 177°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.46
Quantity 5 g
Beilstein 17,II,32
Merck Index 15, 3938
Specific Gravity 0.9225
Specific Rotation -0.3° to +0.3° (20°C, 589nm) (neat)
Solubility Information (in ethanol (70% v/v)) 1:2
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442470 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog Number
Lot NumberA0393363 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,8-Cineole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.