missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals

57.89 € - 159.80 €

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
Synonym amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
PubChem CID 64150
ChEBI CHEBI:2619
IUPAC Name adamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10645972
View Documents
Thermo Scientific Acros
151670250
25 g
57.89 €
25g
Please to purchase this item. Need a web account? Register with us today!
10571781
View Documents
Thermo Scientific Acros
151671000
100 g
159.80 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Specifications

Chemical Identifiers

665-66-7
187.71
WOLHOYHSEKDWQH-UHFFFAOYSA-N
64150
adamantan-1-amine;hydrochloride
C10H18ClN
MFCD00074723
amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
CHEBI:2619
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifications

665-66-7
100
White
99+%
C10H18ClN
15, 368
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2
187.71
CHEBI:2619
99+%
1-Adamantanamine hydrochloride, 99+%
99
>300.0°C
Authentic
Glass bottle
MFCD00074723
amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
WOLHOYHSEKDWQH-UHFFFAOYSA-N
adamantan-1-amine;hydrochloride
64150
187.71
Fine Crystalline Powder
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

Harmful if swallowed

Acute toxicity (category 4)

missing translation for 'einecsNumber' : 211-560-2

missing translation for 'rtecsNumber' : AU4375000

missing translation for 'tsca' : TSCA

Produktidentifikator
  • 1-Adamantanamine hydrochloride

Signalwort
  • Warning

Gefahrenkategorie
  • Acute toxicity Category 4

Gefahrenhinweis
  • H302-Harmful if swallowed.

Sicherheitshinweis
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt