missing translation for 'onlineSavingsMsg'
Learn More
1-Benzyl-4-piperidone, 98+%, Thermo Scientific Chemicals Product Code.: 11429553

1-Benzyl-4-piperidone, 98+%, Thermo Scientific Chemicals

Product Code. 11429553
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11429553

Brand: Thermo Scientific Alfa Aesar A12390.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

Solubility
Soluble in water (12 g/100ml at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-oneShow MoreShow Less
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

Density 1.059
Boiling Point 248°C
Flash Point >110°C (230°F)
Refractive Index 1.5399
Quantity 25 g
Beilstein 128556
Solubility Information Soluble in water (12g/100ml at 20°C).
Formula Weight 189.26
Percent Purity ≥98%
Chemical Name or Material 1-Benzyl-4-piperidone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252498 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog NumberA12390.22,A12390.14,A12390.36
Lot Number10245003 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog NumberA12390.22,A12390.14,A12390.36
Lot Number10200005 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12390.22,A12390.14,A12390.36
Lot Number10229100 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12390.22,A12390.14,A12390.36
Lot Number10211906 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12390.22,A12390.14,A12390.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Benzyl-4-piperidone, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.