missing translation for 'onlineSavingsMsg'
Learn More
Donepezil hydrochloride, Thermo Scientific Chemicals Product Code.: 15492202

Donepezil hydrochloride, Thermo Scientific Chemicals

Product Code. 15492202
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15492202

Brand: Thermo Scientific Acros 458050010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24H30ClNO3
Molecular Weight (g/mol) 415.96
MDL Number MFCD00881312
InChI Key XWAIAVWHZJNZQQ-UHFFFAOYNA-N
Synonym donepezil hydrochloride, donepezil hcl, aricept, bnag, donepezilhcl, aricept odt, 2-1-benzylpiperidin-4-yl methyl-5,6-dimethoxy-2,3-dihydro-1h-inden-1-one hydrochloride, e 2020 pharmaceutical, eranzShow MoreShow Less
PubChem CID 5741
ChEBI CHEBI:4696
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride
SMILES [H+].[Cl-].COC1=CC2=C(C=C1OC)C(=O)C(CC1CCN(CC3=CC=CC=C3)CC1)C2

Specifications

Melting Point 211.0°C to 212.0°C
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot methanol, insoluble in ethyl acetate and hexane
Formula Weight 415.85
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Donepezil hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455173 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number458050010,458050050
Lot NumberA0395311 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458050010,458050050
Lot NumberA0445181 Certificate TypeCertificate of Analysis Date24/08/2022 Catalog Number458050010,458050050
Lot NumberA0443263 Certificate TypeCertificate of Analysis Date21/06/2022 Catalog Number458050010,458050050
Lot NumberA0439997 Certificate TypeCertificate of Analysis Date23/03/2022 Catalog Number458050010,458050050
compliance-icons
Product Identifier
  • Donepezil hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Donepezil hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Donepezil hydrochloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.