Learn More
1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals
Description
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .

Specifications
Specifications
Melting Point | -8°C |
Density | 1.601 |
Boiling Point | 155°C to 157°C |
Flash Point | 43°C (109°F) |
Odor | Aromatic |
Refractive Index | 1.534 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 1071819 |
Solubility Information | Insoluble in water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10237449 | Certificate of Analysis | 12/04/2022 | A10635.22,A10635.14,A10635.36 |
5005F02W | Certificate of Analysis | 22/11/2021 | A10635.22,A10635.14,A10635.36 |
10138140 | Certificate of Analysis | 22/11/2021 | A10635.22,A10635.14,A10635.36 |
10198737 | Certificate of Analysis | 22/11/2021 | A10635.22,A10635.14,A10635.36 |
10220497 | Certificate of Analysis | 22/11/2021 | A10635.22,A10635.14,A10635.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.