missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals

41.71 € - 527.63 €

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name 3-bromopropylbenzene
SMILES BrCCCC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477956
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14050.22
100 g
41.71 €
100g
Please to purchase this item. Need a web account? Register with us today!
11487956
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14050.36
500 g
133.67 €
500g
Please to purchase this item. Need a web account? Register with us today!
11497956
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14050.0E
2500 g
527.63 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

Solubility
Soluble in water (partly). Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
Specifications

Chemical Identifiers

637-59-2
199.09
XMZQWZJMTBCUFT-UHFFFAOYSA-N
12503
BrCCCC1=CC=CC=C1
C9H11Br
MFCD00000257
3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
3-bromopropylbenzene

Specifications

637-59-2
219°C to 220°C
C9H11Br
MFCD00000257
3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
XMZQWZJMTBCUFT-UHFFFAOYSA-N
3-bromopropylbenzene
12503
98%
1.308
101°C (213°F)
1.546
2205527
Soluble in water (partly). Soluble in chloroform and methanol.
BrCCCC1=CC=CC=C1
199.09
199.09
1-Bromo-3-phenylpropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 211-294-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Bromo-3-phenylpropane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 1-Bromo-3-phenylpropane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only