missing translation for 'onlineSavingsMsg'
Learn More
1-Bromobutane, 98+%, Thermo Scientific Chemicals Product Code.: 11493454

1-Bromobutane, 98+%, Thermo Scientific Chemicals

Product Code. 11493454
500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11493454

Brand: Thermo Scientific Alfa Aesar A10696.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831Show MoreShow Less
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr

Specifications

Melting Point °C to 112°C
Density 1.275 g/mL
Boiling Point 101°C to 102°C
Flash Point 20°C (68°F)
Assay Percent Range >98%
Linear Formula CH3(CH2)3Br
Refractive Index 1.44
Quantity 500 g
UN Number UN1126
Beilstein 1098260
Merck Index 14,1553
Solubility Information Miscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 137.03
Percent Purity ≥98%
Chemical Name or Material 1-Bromobutane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252082 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10252083 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10226499 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10245504 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10242084 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog NumberA10696.0E,A10696.0C,A10696.36
compliance-icons compliance-icons
Product Identifier
  • 1-Bromobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : Butyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromobutane, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.