missing translation for 'onlineSavingsMsg'
Learn More
1-Bromobutane, 99%, Thermo Scientific Chemicals Product Code.: 10509820

1-Bromobutane, 99%, Thermo Scientific Chemicals

Product Code. 10509820
250 mL, Glass bottle
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10509820

Brand: Thermo Scientific Acros 106772500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831Show MoreShow Less
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -112.0°C
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 100.0°C to 104.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3Br
Refractive Index 1.4380 to 1.4404
Quantity 250 mL
Beilstein 01,119
Merck Index 15,1555
Specific Gravity 1.27
Solubility Information Solubility in water: 0.608g/L (30°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene and ccl4
Formula Weight 137.02
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromobutane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465682 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number106770025,106772500,106770100,106770010
Lot NumberA0450763 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number106770025,106772500,106770100,106770010
Lot NumberA0458432 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number106770025,106772500,106770100,106770010
Lot NumberA0448244 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number106770025,106772500,106770100,106770500,106770010
Lot NumberA0397703 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number106770025,106772500,106770100,106770500,106770010
compliance-icons compliance-icons
Product Identifier
  • 1-Bromobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromobutane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.