Learn More
1-Bromoheptane, 98+%, Thermo Scientific Chemicals
43.42 € - 385.92 €
Chemical Identifiers
CAS | 629-04-9 |
---|---|
Molecular Formula | C7H15Br |
Molecular Weight (g/mol) | 179.10 |
MDL Number | MFCD00000273 |
InChI Key | LSXKDWGTSHCFPP-UHFFFAOYSA-N |
Synonym | heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane |
PubChem CID | 12369 |
IUPAC Name | 1-bromoheptane |
SMILES | CCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467916
|
Thermo Scientific Alfa Aesar
A14031.30 |
250 g |
43.42 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457916
|
Thermo Scientific Alfa Aesar
A14031.0B |
1000 g |
109.55 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477916
|
Thermo Scientific Alfa Aesar
A14031.0I |
5000 g |
385.92 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Immiscible with water.
Notes
Flammable and incompatible with oxidizing agents, strong bases.
Chemical Identifiers
629-04-9 | |
179.10 | |
LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
12369 | |
CCCCCCCBr |
C7H15Br | |
MFCD00000273 | |
heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
1-bromoheptane |
Specifications
629-04-9 | |
1.139 | |
60°C (140°F) | |
C7H15Br | |
MFCD00000273 | |
1697160 | |
Immiscible with water. | |
CCCCCCCBr | |
179.10 | |
179.11 | |
1-Bromoheptane |
-58°C | |
179°C to 180°C | |
Recognizable | |
1.4499 | |
UN1993 | |
heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
1-bromoheptane | |
12369 | |
≥98% |
Safety and Handling
GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 211-068-8
missing translation for 'rtecsNumber' : MI8100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.