Learn More
1-Butanol, 99%, Thermo Scientific Chemicals
25.73 € - 53.87 €
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00002964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
Synonym | 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane |
PubChem CID | 263 |
ChEBI | CHEBI:28885 |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367946
|
Thermo Scientific Alfa Aesar
L13171.AE |
100 mL |
25.73 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377946
|
Thermo Scientific Alfa Aesar
L13171.AU |
1000 mL |
30.95 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387946
|
Thermo Scientific Alfa Aesar
L13171.0F |
2500 mL |
53.87 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.
Solubility
Miscible with alcohols, ketones, aldehydes, ethers, aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents, alkali metals and halogens.
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Specifications
7 | |
71-36-3 | |
117°C to 118°C | |
Alcohol-like | |
C4H10O | |
UN1120 | |
14,1540 | |
Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water. | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
99% |
-90°C to -89°C | |
0.81 | |
35°C (95°F) | |
1.399 | |
MFCD00002964 | |
969148 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12 | |
1-Butanol |
Safety and Handling
GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H318-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
missing translation for 'einecsNumber' : 200-751-6
missing translation for 'rtecsNumber' : EO1400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Butanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : n-Butyl alcohol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.