missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)
464.30 € - 710.18 €
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00002964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
Synonym | 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane |
PubChem CID | 263 |
ChEBI | CHEBI:28885 |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15693880
|
Honeywell-Riedel-de Haen
33065-6X1L |
6 x 1 L | Poly Bottle |
464.30 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15683880
|
Honeywell-Riedel-de Haen
33065-4X2.5L |
4 x 2.5 L | Not available |
710.18 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Specifications
71-36-3 | |
0.81g/mL at 25°C | |
35°C (95°F) | |
n20/D 1.399 | |
CH3(CH2)3OH | |
UN1120 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12g/mol | |
ACS/puriss. p.a. |
Colorless | |
0.112 | |
649°F | |
C4H10O | |
MFCD00002964 | |
969148 | |
66g/L in water at 20°C, pH 7 | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
≥99.5% (GC) | |
1-Butanol, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
H226-H302-H315-H318-H335-H336
Danger
missing translation for 'dotInformation' : Class 3 - PG 3 - Butanols
missing translation for 'einecsNumber' : 200-751-6
missing translation for 'rtecsNumber' : EO1400000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Butanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title