missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid

Specifications
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Melting Point | -71.0°C |
Color | Orange-Yellow |
Density | 1.2077g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Merck Index | 15, 1584 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466807 | Certificate of Analysis | 16/10/2024 | 354211000,354210250,354210050 |
A0450450 | Certificate of Analysis | 02/05/2023 | 354211000,354210250,354210050 |
A0394522 | Certificate of Analysis | 14/12/2022 | 354211000,354210250,354210050 |
A0400929 | Certificate of Analysis | 14/12/2022 | 354211000,354210250,354210050 |
A0406743 | Certificate of Analysis | 13/12/2022 | 354211000,354210250,354210050 |
Safety and Handling

Product Identifier
- 1-Butyl-3-methylimidazolium tetrafluoroborate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction