missing translation for 'onlineSavingsMsg'
Learn More
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals Product Code.: 10551282

1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Product Code. 10551282
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10551282

Brand: Thermo Scientific Acros 354210050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborateShow MoreShow Less
PubChem CID 2734178
IUPAC Name 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -71.0°C
Color Orange-Yellow
Density 1.2077g/mL
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1584
Specific Gravity 1.2077
Viscosity 118.3 mPa.s (25°C)
Formula Weight 226.02
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium tetrafluoroborate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466807 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number354211000,354210250,354210050
Lot NumberA0450450 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number354211000,354210250,354210050
Lot NumberA0394522 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number354211000,354210250,354210050
Lot NumberA0400929 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number354211000,354210250,354210050
Lot NumberA0406743 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number354211000,354210250,354210050
compliance-icons
Product Identifier
  • 1-Butyl-3-methylimidazolium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.