Learn More
1-Chlorophthalazine, 97%, Thermo Scientific Chemicals
Description
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.

Specifications
Specifications
Melting Point | 109°C to 112°C |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. Soluble in dichloromethane,ethanol and methanol. |
Formula Weight | 164.59 |
Percent Purity | 97% |
Chemical Name or Material | 1-Chlorophthalazine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10240542 | Certificate of Analysis | 20/02/2024 | H27143.09,H27143.03 |
10223516 | Certificate of Analysis | 07/12/2021 | H27143.09,H27143.03 |
F21X025 | Certificate of Analysis | 07/12/2021 | H27143.09,H27143.03 |
10159233 | Certificate of Analysis | 07/12/2021 | H27143.09,H27143.03 |
10135900 | Certificate of Analysis | 07/12/2021 | H27143.09,H27143.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.