missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals

Product Code. 11334098
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 x 500 mL

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorhexidine digluconate, 20% w/v aq. Soln is used as an oral antiseptic agent and anti-microbial agent. It is an active ingredient in germicidal mouthwash, pet shampoo, surgical lubricants and wipes and hand soap. It is utilized in the study to know the role of essential oils for improving skin antisepsis. It plays a vital role as disinfectants and preservative in pharmaceutical products. It is also used as an additive in cosmetics such as creams, toothpaste and deodorants.

Solubility
Miscible with water, ethanol and acetone.

Notes
Store in a cool place. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C34H54Cl2N10O14
Molecular Weight (g/mol) 897.76
MDL Number MFCD00083599
InChI Key YZIYKJHYYHPJIB-UUPCJSQJSA-N
Synonym peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate
PubChem CID 60148207
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Color Colorless
Density 1.06 g/mL
Assay Percent Range 20% w/v aq. soln.
Quantity 100 mL
Sensitivity Light sensitive
Formula Weight 897.76
Concentration or Composition (by Analyte or Components) 20% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Chlorhexidine digluconate, Non-sterile
compliance-icons
Product Identifier
  • Chlorhexidine digluconate, 20% w/v aqueous solution, non-sterile
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Chlorhexidine digluconate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.