missing translation for 'onlineSavingsMsg'
Learn More
(1-Formylcyclopropyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™ Product Code.: 15560554

(1-Formylcyclopropyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™

Product Code. 15560554
1 g
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15560554

Brand: Thermo Scientific Acros 449950010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 107259-06-3
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C9H15NO3
Synonym tert-butyl 1-formylcyclopropyl carbamate, tert-butyl 1-formylcyclopropylcarbamate, 1-formyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1-formylcyclopropyl carbamate, pubchem17629, tert-butyl 1-formylcyclopropyl-carbamate, n-boc-1-amino-1-cyclopropanecarboxaldehyde, tert-butyl l-formylcyclopropyl-carbamate
InChI Key ACMHCEYCNRURST-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1(CC1)C=O
IUPAC Name tert-butyl N-(1-formylcyclopropyl)carbamate
Molecular Weight (g/mol) 185.22
Quantity 1 g
PubChem CID 10352385
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (1-Formylcyclopropyl)carbamic acid tert-butyl ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0445114 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number
Lot NumberA0363430 Certificate TypeCertificate of Analysis Date28/08/2021 Catalog Number
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1-Formylcyclopropyl)carbamic acid tert-butyl ester
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1-Formylcyclopropyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.