missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals

62.11 € - 2702.45 €

Chemical Identifiers

CAS 207300-90-1
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
PubChem CID 23687711
IUPAC Name sodium;heptane-1-sulfonate;hydrate
SMILES CCCCCCCS(=O)(=O)[O-].O.[Na+]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10516982
View Documents
Thermo Scientific Acros
411270050
5 g Glass bottle
62.11 €
5g
Please to purchase this item. Need a web account? Register with us today!
10107330
View Documents
Thermo Scientific Acros
411270250
25 g Glass Bottle
239.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
10403551
View Documents
Thermo Scientific Acros
411271000
100 g Glass Bottle
750.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
11904571
View Documents
Thermo Scientific Acros
411275000
500 g Glass bottle
2702.45 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
Specifications

Chemical Identifiers

207300-90-1
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
sodium;heptane-1-sulfonate;hydrate
XWZCREJRXRKIRQ-UHFFFAOYSA-M
23687711
CCCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

207300-90-1
>300.0°C
7.4% to 9% (100°C, 3 hrs) (vacuum)
C7H15NaO3S·H2O
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
XWZCREJRXRKIRQ-UHFFFAOYSA-M
sodium;heptane-1-sulfonate;hydrate
23687711
Powder or Flakes
(2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
White
Authentic
CH3(CH2)6SO3Na·H2O
(10% w/v in water) Clear colorless
CCCCCCCS(=O)(=O)[O-].O.[Na+]
220.27
220.27
1-Heptanesulfonic acid sodium salt monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • 1-Heptanesulfonic acid, sodium salt monohydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 1-Heptanesulfonic acid, sodium salt monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only