missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™ Product Code.: 10235554

1-Methyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10235554
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10235554

Brand: Thermo Scientific Maybridge CC48302CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 84547-60-4
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD03419367
InChI Key JCZIPPIUHKRMOM-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-carbonyl chloride, 1h-pyrazole-3-carbonyl chloride, 1-methyl, 1-methyl-3-pyrazolecarbonyl chloride, art-chem-bb b006627, 1h-pyrazole-3-carbonylchloride, 1-methyl, 1h-pyrazole-3-carbonyl chloride, 1-methyl-9ciShow MoreShow Less
PubChem CID 12845045
IUPAC Name 1-methylpyrazole-3-carbonyl chloride
SMILES CN1C=CC(=N1)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-3-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.