missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-3-carboxylic acid, 95%, Thermo Scientific™

41.90 € - 122.00 €

Chemical Identifiers

CAS 25016-20-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464254
InChI Key YBFIKNNFQIBIQZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
PubChem CID 573176
ChEBI CHEBI:74738
IUPAC Name 1-methylpyrazole-3-carboxylic acid
SMILES CN1C=CC(=N1)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10746905
View Documents
Thermo Scientific Maybridge
CC48301CB
250 mg Amber Glass Bottle
41.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10226753
View Documents
Thermo Scientific Maybridge
CC48301DA
1 g Amber Glass Bottle
122.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

25016-20-0
126.12
YBFIKNNFQIBIQZ-UHFFFAOYSA-N
573176
1-methylpyrazole-3-carboxylic acid
C5H6N2O2
MFCD00464254
1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
CHEBI:74738
CN1C=CC(=N1)C(O)=O

Specifications

25016-20-0
100.0
C5H6N2O2
1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
CN1C=CC(=N1)C(O)=O
126.12
CHEBI:74738
95%
94.5
95%
MFCD00464254
YBFIKNNFQIBIQZ-UHFFFAOYSA-N
1-methylpyrazole-3-carboxylic acid
573176
126.11
1-Methyl-1H-pyrazole-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.