missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals Product Code.: 11450630

1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals

Product Code. 11450630
500 mL
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11450630

Brand: Thermo Scientific Alfa Aesar 043894.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS Number-872-50-4, 4L, 106420, 204 deg.C, Colorless to Yellow, 1.033 g/mL, 212-828-1, 86 deg.C (187 deg.F), 99.13, ACS Reagent, H227-H315-H319-H335-H360D-H373, P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c, -24 deg.C

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24°C
Color Colorless to Yellow
Density 1.033 g/mL
Boiling Point 204°C
Quantity 500 mL
Flash Point 86°C (187°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.4705
Beilstein 106420
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Grade ACS Reagent
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY26K061 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog Number043894.AP,043894.M1,043894.M6,043894.K2,043894.K7
Lot NumberT11K023 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number043894.AP,043894.M1,043894.M6,043894.K2,043894.K7
Lot NumberW09K021 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number043894.AP,043894.M1,043894.M6,043894.K2,043894.K7
Lot NumberZ06J024 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number043894.AP,043894.M1,043894.M6,043894.K2,043894.K7
Lot NumberT28J040 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number043894.AP,043894.M1,043894.M6,043894.K2,043894.K7
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone, ACS grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-methyl-2-pyrrolidone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.