missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, CHROMASOLV™ Plus, for HPLC, ≥99%, Honeywell Riedel-de Haën™
CHROMASOLV™ Plus, for HPLC,≥99%
187.74 € - 1115.32 €
Chemical Identifiers
CAS | 872-50-4 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.133 |
MDL Number | MFCD00003193 |
InChI Key | SECXISVLQFMRJM-UHFFFAOYSA-N |
Synonym | 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl |
PubChem CID | 13387 |
ChEBI | CHEBI:7307 |
IUPAC Name | 1-methylpyrrolidin-2-one |
SMILES | CN1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15662840
|
Honeywell-Chromasolv
270458-1L |
1 L | Glass Bottle |
187.74 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15672840
|
Honeywell-Chromasolv
270458-2.5L |
2.5 L | Glass Bottle |
450.14 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15682840
|
Honeywell-Chromasolv
270458-4X4L |
4 x 4 L | Glass Bottle | N/A | |||||
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
872-50-4 | |
99.133 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
13387 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
MFCD00003193 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
CHEBI:7307 | |
CN1CCCC1=O |
Specifications
-24°C (literature) | |
872-50-4 | |
1.028 g/mL at 25 °C(lit.) | |
91°C | |
C5H9NO | |
106420 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
13387 | |
99.13 | |
HPLC |
max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 325 nm, max. 0.50 at 300 nm, max. 1.0 at 285 nm | |
100.0000% | |
204°C | |
C5H9NO | |
MFCD00003193 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
CN1CCCC1=O | |
99.133 | |
CHEBI:7307 | |
>99% | |
1-Methyl-2-pyrrolidinone, CHROMASOLV™ Plus, for HPLC, ≥99% |
Safety and Handling
missing translation for 'shelfLife' : 1980 days from date of manufacture
missing translation for 'einecsNumber' : 212-828-1
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1-Methyl-2-pyrrolidinone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H360D-May damage the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N-Methyl-2-pyrrolidone
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title