missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbenzimidazole-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

102.00 € - 388.94 €

Chemical Identifiers

CAS 3012-80-4
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00142655
InChI Key SIRPHJCQZYVEES-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzoimidazole-2-carbaldehyde, 1-methyl-1h-benzimidazole-2-carboxaldehyde, 1-methyl-1h-benzimidazole-2-carbaldehyde, 1-methyl-1h-benzo d imidazole-2-carbaldehyde, 1-methyl-2-formylbenzimidazole, 1h-benzimidazole-2-carboxaldehyde, 1-methyl, 1-methyl-1h-1,3-benzodiazole-2-carbaldehyde, 1-methyl-1,3-benzodiazole-2-carbaldehyde, 1-methyl-2-benzimidazolecarboxaldehyde, pubchem16250
PubChem CID 762084
IUPAC Name 1-methylbenzimidazole-2-carbaldehyde
SMILES CN1C(C=O)=NC2=CC=CC=C12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11461627
View Documents
Thermo Scientific Alfa Aesar
B25601.03
1 g
102.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11471627
View Documents
Thermo Scientific Alfa Aesar
B25601.06
5 g
388.94 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3012-80-4
160.18
SIRPHJCQZYVEES-UHFFFAOYSA-N
762084
CN1C(C=O)=NC2=CC=CC=C12
C9H8N2O
MFCD00142655
1-methyl-1h-benzoimidazole-2-carbaldehyde, 1-methyl-1h-benzimidazole-2-carboxaldehyde, 1-methyl-1h-benzimidazole-2-carbaldehyde, 1-methyl-1h-benzo d imidazole-2-carbaldehyde, 1-methyl-2-formylbenzimidazole, 1h-benzimidazole-2-carboxaldehyde, 1-methyl, 1-methyl-1h-1,3-benzodiazole-2-carbaldehyde, 1-methyl-1,3-benzodiazole-2-carbaldehyde, 1-methyl-2-benzimidazolecarboxaldehyde, pubchem16250
1-methylbenzimidazole-2-carbaldehyde

Specifications

3012-80-4
C9H8N2O
Air Sensitive
SIRPHJCQZYVEES-UHFFFAOYSA-N
1-methylbenzimidazole-2-carbaldehyde
762084
≥98%
120°C to 122°C
MFCD00142655
1-methyl-1h-benzoimidazole-2-carbaldehyde, 1-methyl-1h-benzimidazole-2-carboxaldehyde, 1-methyl-1h-benzimidazole-2-carbaldehyde, 1-methyl-1h-benzo d imidazole-2-carbaldehyde, 1-methyl-2-formylbenzimidazole, 1h-benzimidazole-2-carboxaldehyde, 1-methyl, 1-methyl-1h-1,3-benzodiazole-2-carbaldehyde, 1-methyl-1,3-benzodiazole-2-carbaldehyde, 1-methyl-2-benzimidazolecarboxaldehyde, pubchem16250
CN1C(C=O)=NC2=CC=CC=C12
160.18
160.17
1-Methylbenzimidazole-2-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only