missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylamine hydrochloride, Thermo Scientific Chemicals

42.41 € - 153.77 €

Chemical Identifiers

CAS 1838-08-0
Molecular Formula C18H40ClN
Molecular Weight (g/mol) 305.975
MDL Number MFCD00042018
InChI Key RNYJXPUAFDFIQJ-UHFFFAOYSA-N
Synonym octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
PubChem CID 519552
IUPAC Name octadecan-1-amine;hydrochloride
SMILES CCCCCCCCCCCCCCCCCCN.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364088
SDS View Documents
Thermo Scientific Alfa Aesar
041378.06
5 g
42.41 €
5g
Please to purchase this item. Need a web account? Register with us today!
11374088
SDS View Documents
Thermo Scientific Alfa Aesar
041378.14
25 g
153.77 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Octadecylamine hydrochloride is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1838-08-0
305.975
RNYJXPUAFDFIQJ-UHFFFAOYSA-N
519552
CCCCCCCCCCCCCCCCCCN.Cl
C18H40ClN
MFCD00042018
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
octadecan-1-amine;hydrochloride

Specifications

1838-08-0
C18H40ClN
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
RNYJXPUAFDFIQJ-UHFFFAOYSA-N
octadecan-1-amine;hydrochloride
519552
Powder
160°C
MFCD00042018
Soluble in hot methanol.
CCCCCCCCCCCCCCCCCCN.Cl
305.975
305.97
1-Octadecylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 217-410-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.