Learn More
1-Octylboronic acid, 97%, Thermo Scientific Chemicals
25.93 € - 289.44 €
Chemical Identifiers
CAS | 28741-08-4 |
---|---|
Molecular Formula | C8H19BO2 |
Molecular Weight (g/mol) | 158.048 |
MDL Number | MFCD01074560 |
InChI Key | GKFRVXOKPXCXAK-UHFFFAOYSA-N |
Synonym | n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid |
PubChem CID | 5195177 |
IUPAC Name | octylboronic acid |
SMILES | B(CCCCCCCC)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334346
|
Thermo Scientific Alfa Aesar
L19964.03 |
1 g |
25.93 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344346
|
Thermo Scientific Alfa Aesar
L19964.06 |
5 g |
86.73 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354346
|
Thermo Scientific Alfa Aesar
L19964.14 |
25 g |
289.44 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Octylboronic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octylboronic acid is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
28741-08-4 | |
158.048 | |
GKFRVXOKPXCXAK-UHFFFAOYSA-N | |
5195177 | |
B(CCCCCCCC)(O)O |
C8H19BO2 | |
MFCD01074560 | |
n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid | |
octylboronic acid |
Specifications
28741-08-4 | |
C8H19BO2 | |
1739759 | |
n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid | |
GKFRVXOKPXCXAK-UHFFFAOYSA-N | |
octylboronic acid | |
5195177 | |
97% |
81°C to 85°C | |
MFCD01074560 | |
Air Sensitive | |
Insoluble in water. | |
B(CCCCCCCC)(O)O | |
158.048 | |
158.05 | |
1-Octylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.