missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, puriss. p.a., Honeywell
BIOPHARMA

100.03 € - 2916.72 €

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15693980
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
33538-1L
1 L
100.03 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15603990
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
33538-2.5L
2.5 L
219.92 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15683980
View Documents
Honeywell Riedel-de Haën
33538-160KG
160 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO

Specifications

71-23-8
0.137
700°F
CH3CH2CH2OH
14.9 mmHg (20°C), 10 mmHg (147°C)
1098242
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1g/mol
puriss. p.a.
0.804g/mL at 25°C
22°C (71.6°F)
C3H8O
MFCD00002941
UN1274
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.10
CHEBI:28831
≥99.5% (GC)
1-Propanol
Videos
Safety and Handling

Safety and Handling

P210-P261-P280-P305 + P351 + P338

Danger

missing translation for 'einecsNumber' : 200-746-9

Product Identifier
  • 1-Propanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : propan-1-ol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.