missing translation for 'onlineSavingsMsg'
Learn More
1-Propanol, Thermo Scientific Chemicals Product Code.: 12397163

1-Propanol, Thermo Scientific Chemicals

Product Code. 12397163
500 mL
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 12397163

Brand: Thermo Scientific Acros 445775000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcoholShow MoreShow Less
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127.0°C
Density 0.8000g/mL
Boiling Point 97.0°C
Quantity 500 mL
Flash Point 15°C
Refractive Index 1.3840 to 1.3860
Specific Gravity 0.8
Linear Formula CH3CH2CH2OH
Viscosity 2.2 mPa.s (20°C)
Water 0.2% max.
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Formula Weight 60.1
Percent Purity ≥99.5% (GC) 0.01% max. (Ethanol) (GC) 0.01% max. (Methanol) (GC) 0.05% max. (Isopropyl alcohol) (GC)
Grade ACS Reagent
Chemical Name or Material 1-Propanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466333 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number445770010,445775000
Lot NumberA0437648 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number445770010,445775000
Lot NumberA0428512 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number445770010,445775000
Lot NumberA0460681 Certificate TypeCertificate of Analysis Date01/03/2024 Catalog Number445770010,445775000
Lot NumberA0453944 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number445770010,445775000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : n-Propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Propanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.