missing translation for 'onlineSavingsMsg'
Learn More

1-Trimethylsilyl-1-pentyne, 98%, Thermo Scientific Chemicals

33.87 € - 360.80 €

Chemical Identifiers

CAS 18270-17-2
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00077890
InChI Key CABCDUQQPBAHEE-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne
PubChem CID 2760860
SMILES CCCC#C[Si](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15450218
SDS View Documents
Thermo Scientific Alfa Aesar
H53423.06
5 g
33.87 €
5g
Please to purchase this item. Need a web account? Register with us today!
15460218
SDS View Documents
Thermo Scientific Alfa Aesar
H53423.14
25 g
104.52 €
25g
Please to purchase this item. Need a web account? Register with us today!
15470218
SDS View Documents
Thermo Scientific Alfa Aesar
H53423.22
100 g
360.80 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18270-17-2
140.30
CABCDUQQPBAHEE-UHFFFAOYSA-N
2760860
C8H16Si
MFCD00077890
1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne
CCCC#C[Si](C)(C)C

Specifications

135°C
C8H16Si
MFCD00077890
1744218
1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne
CCCC#C[Si](C)(C)C
140.30
140.3
1.425
1-Trimethylsilyl-1-pentyne
18270-17-2
CH3CH2CH2C≡CSi(CH3)3
UN1993
23°C (73°F)
CABCDUQQPBAHEE-UHFFFAOYSA-N
trimethyl(pent-1-yn-1-yl)silane
2760860
98%
0.765
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.