Learn More
Masterflex™ Stopcocks with Luer Connections
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
Brand:  Masterflex™ 30600-01
 View more versions of this product
Product Code. 11772643
  • 50.00 € / Pack of 10
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
  • Handle rotates 90°
  • PC body and HDPE stem
Specifications
Specifications
Stopcock
Polycarbonate, HDPE
Polycarbonate and HDPE
Male Luer Slip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Stopcocks with Luer Connections > Luer, Male slip