Learn More
Masterflex™ Stopcocks with Luer Connections
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
50.00 € - 54.70 €
Specifications
Type | Stopcock |
---|---|
Material | Polycarbonate, HDPE |
Plug Material | Polycarbonate and HDPE |
Description
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
- Handle rotates 90°
- PC body and HDPE stem
Specifications
Stopcock | |
Polycarbonate, HDPE | |
Polycarbonate and HDPE |