Learn More
Thermo Scientific™ 11 mm Glass Crimp Top Vials
Use these 11 mm clear glass vials with snap caps or aluminum crimp seal closures.
16.10 € - 932.00 €
Product Code | Brand | Volume (Metric) | Type | Base Style | Marking Spot | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Type | Base Style | Marking Spot | Price | Quantity & Availability | |||||
10147404
|
Thermo Scientific™
60180-502 |
2 mL | Crimp Top Vial | Flat Bottom | Yes | N/A | ||||||
13171703
|
Thermo Scientific™
C4012-1 |
2 mL | Crimp Top Vial | Flat Bottom | No | N/A | ||||||
11915713
|
Thermo Scientific™
C4011-1 |
2 mL | Crimp Top Vial | Flat Bottom | No | N/A | ||||||
12453718
|
Thermo Scientific™
C4011-10 |
650 μL | Crimp Top Vial | Conical Base | No | N/A | ||||||
12410068
|
Thermo Scientific™
C4011-LV1 |
500 μL | Snap-It Vial | Fused Conical | No |
108.99 €
Pack of 100 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
10332825
|
Thermo Scientific™
C4011-LV1W |
500 μL | Snap-It Vial | Fused Conical | Yes | N/A | ||||||
12420048
|
Thermo Scientific™
C4011-9 |
1.7 mL | Crimp Top Vial | High Recovery | No | N/A | ||||||
12440068
|
Thermo Scientific™
C4011-S1 |
2 mL | Crimp Top Vial | Flat Bottom | No | N/A | ||||||
12450068
|
Thermo Scientific™
C4011-S1W |
2 mL | Crimp Top Vial | Flat Bottom | Yes | N/A | ||||||
13131703
|
Thermo Scientific™
C4011-S5 |
2 mL | Snap-It Vial | Flat Bottom | No | N/A | ||||||
12400078
|
Thermo Scientific™
C4011-S5W |
2 mL | Snap-It Vial | Flat Bottom | Yes | N/A | ||||||
11931563
|
Thermo Scientific™
C4011-5 |
2 mL | Snap-It Vial | Flat Bottom | No | N/A | ||||||
10599375
|
Thermo Scientific™
C4011-5W |
2 mL | Snap-It Vial | Flat Bottom | Yes | N/A | ||||||
10189184
|
Thermo Scientific™
C4011-4 |
1.7 mL | Snap-It Vial | High Recovery | No | N/A | ||||||
12470068
|
Thermo Scientific™
C4011-S4 |
1.5 mL | Snap-It Vial | High Recovery | No | N/A | ||||||
10360951
|
Thermo Scientific™
09-CTV |
1.0 mL | Crimp Top Vial | Conical | No | N/A | ||||||
11523780
|
Thermo Scientific™
2.5-CV |
2.5 mL | Crimp Top Vial | Flat Bottom | No | N/A | ||||||
11563840
|
Thermo Scientific™
60180-710 |
2.0 mL | Crimp/Snap Vial | Flat Bottom | No | N/A | ||||||
12089143
|
Thermo Scientific™
2-CRV |
2.0 mL | Crimp Top Vial | Round Bottom | Yes | N/A | ||||||
10207234
|
Thermo Scientific™
2-CV |
2.0 mL | Crimp Top Vial | Flat Bottom | Yes | N/A | ||||||
11563760
|
Thermo Scientific™
4-CV |
4.0 mL | Crimp Top Vial | Flat Bottom | No | N/A | ||||||
11543780
|
Thermo Scientific™
09-FIV |
0.9 mL | Crimp Top Vial | Insert Vial | No | N/A | ||||||
10798345
|
Thermo Scientific™
03-FIV |
300 μL | Crimp Top Vial | Insert Vial | Yes | N/A | ||||||
10583703
|
Thermo Scientific™
1.5-HRRV(S) |
1.5 mL | Snap Cap Vial | High Recovery | No |
258.00 €
Pack of 100 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
11523790
|
Thermo Scientific™
1.5-HRRV |
1.5 mL | Snap Cap Vial | High Recovery | No | N/A | ||||||
12396818
|
Thermo Scientific™
1.2-UHRRV |
1.2 mL | Snap Cap Vial | Mandrel Base | No | N/A | ||||||
11387371
|
Thermo Scientific™
C4011-9TR |
1.5 mL | Snap-It Vial | Deep Well Base | No | N/A | ||||||
11513790
|
Thermo Scientific™
1.5-HRCV |
1.5 mL | Crimp Top Vial | High Recovery | No | N/A | ||||||
11553690
|
Thermo Scientific™
03-FIRV |
300 μL | Snap Cap Vial | Fused Conical | Yes | N/A | ||||||
11543730
|
Thermo Scientific™
2-RV |
2.0 mL | Snap Cap Vial | Flat Bottom | Yes | N/A | ||||||
12400088
|
Thermo Scientific™
C4012-15 |
475 μL | Snap-It Vial | Insert Vial | No | N/A | ||||||
12410088
|
Thermo Scientific™
C4012-1W |
2 mL | Crimp Top Vial | Flat Bottom | Yes | N/A | ||||||
12480078
|
Thermo Scientific™
C4012-10 |
425 μL | Crimp/Snap Vial | Narrow Conical | No | N/A | ||||||
11981513
|
Thermo Scientific™
C4011-1W |
2 mL | Crimp Top Vial | Flat Bottom | Yes |
17.85 €
Pack of 100 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
12163771
|
Thermo Scientific™
C4011-1B |
2 mL | Crimp Top ColorBand Vial | Flat Bottom | Blue | N/A | ||||||
12173771
|
Thermo Scientific™
C4011-1G |
2 mL | Crimp Top ColorBand Vial | Flat Bottom | Green | N/A | ||||||
12183771
|
Thermo Scientific™
C4011-1R |
2 mL | Crimp Top ColorBand Vial | Flat Bottom | Red | N/A | ||||||
12193771
|
Thermo Scientific™
C4011-1Y |
2 mL | Crimp Top ColorBand Vial | Flat Bottom | Yellow | N/A | ||||||
10008574
|
Thermo Scientific™
C4011-V1 |
1.5 mL | Crimp Top Vial | High Recovery | No | N/A | ||||||
Description
Use Thermo Scientific™ 11 mm Glass Crimp Top Vials with aluminum crimp seal closures. These high-quality 11 mm glass vials are 2 mL, 12 x 32 mm and fit most brands of autosamplers. Made of superior quality 33 expansion borosilicate clear (Type 1, Class A), they meet all requirements of US, EU, JPN Pharmacopia. Choose from Thermo Scientific™, Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
Choose the right vial for your needs
- Easy-to-fill wide neck opening vials require 6 mm diameter micro-inserts
- Identify sample and volume with graduated, write-on patch provided on certain vials
- Get maximum sample extraction without need for separate inserts by using Microsampling and High Recovery Vials
- Obtain optimal recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional)
Select the insert for your sample
- Polyspring inserts are self-aligning and provide a cushion against needle contact
- Precision point inserts minimize residual sample loss
- Pulled point inserts are an economical choice for noncritical applications
Autosampler compatibility*
Compatible with most HPLC and GC autosamplers
*See the Columns and Consumables Catalog for list of autosampler compatibility.
Specifications
11 mm |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.