Learn More
Axygen™ 1250μL Universal Pipette Tips
Matrix compatible tips in clear color
Brand: Axygen™ MTX-1250-C
This item is not returnable.
View return policy
Description
- Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
- Maxymum Recovery™ tips allows complete retrieval of precious samples
- Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
- Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
- Volume range: 15 to 1250μL
- Available in bulk or racked formats
Specifications
1250 μL | |
Clear | |
RNase, DNase-free | |
Matrix™ Impact™ Pipette |
Non-sterile | |
Autoclavable | |
Bulk | |
Polypropylene |