missing translation for 'onlineSavingsMsg'
Learn More

Labcon 15 mL ViewPoint™ Centrifuge Thermochromic Tubes
GREENER_CHOICE

Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C

Brand:  Labcon 3210-325-006-9

 View more versions of this product

Product Code. 16619842

  • 53.90 € / Pack of 50
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Real-time temperature signaling
  • Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
  • With attached flat screw caps
  • Black graduations and white writing surface
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
  • Polypropylene USP Class VI resin
  • Centrifuging to 12,500 x G
  • Freezable to -80°C
  • Made in USA
Specifications

Specifications

ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity
Clear to Blue color change at 0°C to 4°C
Single-use
10/IntegraPack™ Bag
10/IntegraPack™ Bag
ViewPoint™
Autoclavable
17 mm
Screw Cap with Integral Molded Elastomeric Seal
17 mm
Conical Centrifuge Tube
Purple
15 mL
Polypropylene USP Class VI Resin
10 Tubes / IntegraPack™ bag/5 bags / Pk./10 Pk. / case.
Sterile
Room Temperature
118 mm
Conical
Yes
2 to 15 in 0.5 increments
12,500 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers